[2-(4,5-Dichloro-3-methyl-1H-pyrazol-1-yl)butyl]amine

Supplier: Matrix Scientific

080382-500MG 080382-1G 080382-5G
102669-374EA 387.69 USD
102669-374 102669-376 102669-378
[2-(4,5-Dichloro-3-methyl-1H-pyrazol-1-yl)butyl]amine
[2-(4,5-Dichloro-3-methyl-1H-pyrazol-1-yl)butyl]amine

Hazard: Irritant

Formula: C₈H₁₃Cl₂N₃
MDL Number: MFCD10699062
CAS Number: 1177292-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR