{[(1-Ethyl-1H-pyrazol-3-yl)methyl]thio}acetic acid

Supplier: Matrix Scientific

080379-500MG 080379-1G
102669-358EA 336.92 USD
102669-358 102669-360
{[(1-Ethyl-1H-pyrazol-3-yl)methyl]thio}acetic acid
{[(1-Ethyl-1H-pyrazol-3-yl)methyl]thio}acetic acid

Hazard: Irritant

Formula: C₈H₁₂N₂O₂S
MDL Number: MFCD09859489
CAS Number: 1170645-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR