[3-(4-Chloro-3-methyl-1H-pyrazol-1-yl)propyl]amine
Supplier: Matrix Scientific
080377-500MG
080377-1G
102669-350EA
516.92
USD
102669-350
102669-352
[3-(4-Chloro-3-methyl-1H-pyrazol-1-yl)propyl]amine
[3-(4-Chloro-3-methyl-1H-pyrazol-1-yl)propyl]amine
Hazard: Irritant
Formula:
C₇H₁₂ClN₃ |
MDL Number:
MFCD03419758 CAS Number: 951235-35-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...