N-(1-Isopropyl-5-methyl-1H-pyrazol-3-yl)acetamide
Supplier: Matrix Scientific
080373-500MG
080373-1G
080373-5G
102669-328EA
516.92
USD
102669-328
102669-330
102669-332
N-(1-Isopropyl-5-methyl-1H-pyrazol-3-yl)acetamide
N-(1-Isopropyl-5-methyl-1H-pyrazol-3-yl)acetamide
Hazard: Irritant
Formula:
C₉H₁₅N₃O |
MDL Number:
MFCD22374855 CAS Number: 1379811-29-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...