3-[(3-Methoxy-4-nitro-1H-pyrazol-1-yl)methyl]-6,7,8,9-tetrahydro-5H-[1,2,4]triazolo[4,3-a]azepine

Supplier: Matrix Scientific

080364-500MG 080364-1G
102669-292EA 387.69 USD
102669-292 102669-294
3-[(3-Methoxy-4-nitro-1H-pyrazol-1-yl)methyl]-6,7,8,9-tetrahydro-5H-[1,2,4]triazolo[4,3-a]azepine
3-[(3-Methoxy-4-nitro-1H-pyrazol-1-yl)methyl]-6,7,8,9-tetrahydro-5H-[1,2,4]triazolo[4,3-a]azepine

Hazard: Irritant

Formula: C₁₂H₁₆N₆O₃
MDL Number: MFCD16345369
CAS Number: 1245772-20-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR