3-[1-(3-Methyl-4-nitro-1H-pyrazol-1-yl)ethyl]-6,7,8,9-tetrahydro-5H-[1,2,4]triazolo[4,3-a]azepine

Supplier: Matrix Scientific

080362-500MG 080362-1G
102669-284EA 387.69 USD
102669-284 102669-286
3-[1-(3-Methyl-4-nitro-1H-pyrazol-1-yl)ethyl]-6,7,8,9-tetrahydro-5H-[1,2,4]triazolo[4,3-a]azepine
3-[1-(3-Methyl-4-nitro-1H-pyrazol-1-yl)ethyl]-6,7,8,9-tetrahydro-5H-[1,2,4]triazolo[4,3-a]azepine

Hazard: Irritant

Formula: C₁₃H₁₈N₆O₂
MDL Number: MFCD16345368
CAS Number: 1245771-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR