1-(N,N-Dimethylglycyl)-3-(trifluoromethyl)-1H-pyrazole-5-carboxylic acid
Supplier: Matrix Scientific
080360-500MG
080360-1G
102669-274EA
516.92
USD
102669-274
102669-276
1-(N,N-Dimethylglycyl)-3-(trifluoromethyl)-1H-pyrazole-5-carboxylic acid
1-(N,N-Dimethylglycyl)-3-(trifluoromethyl)-1H-pyrazole-5-carboxylic acid
Hazard: Irritant
Formula:
C₉H₁₀F₃N₃O₃ |
MDL Number:
MFCD22374854 CAS Number: 1379811-72-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...