{3-[(2,4-Dimethylphenoxy)methyl]phenyl}amine hydrochloride

Supplier: Matrix Scientific

080358-500MG 080358-1G
102669-266EA 387.69 USD
102669-266 102669-268
{3-[(2,4-Dimethylphenoxy)methyl]phenyl}amine hydrochloride
{3-[(2,4-Dimethylphenoxy)methyl]phenyl}amine hydrochloride

Hazard: Irritant

Formula: C₁₅H₁₈ClNO
MDL Number: MFCD22421976
CAS Number: 1417570-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR