{3-[(4-Chlorophenoxy)methyl]phenyl}amine

Supplier: Matrix Scientific

080357-500MG 080357-1G
102669-262EA 387.69 USD
102669-262 102669-264
{3-[(4-Chlorophenoxy)methyl]phenyl}amine
{3-[(4-Chlorophenoxy)methyl]phenyl}amine

Hazard: Irritant

Formula: C₁₃H₁₂ClNO
MDL Number: MFCD09816732
CAS Number: 1016681-15-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR