1-(Difluoromethyl)-1H-pyrazole-5-carbohydrazide
Supplier: Matrix Scientific
080351-500MG
080351-1G
102669-234EA
646.15
USD
102669-234
102669-236
1-(Difluoromethyl)-1H-pyrazole-5-carbohydrazide
1-(Difluoromethyl)-1H-pyrazole-5-carbohydrazide
Hazard: Irritant
Formula:
C₅H₆F₂N₄O |
MDL Number:
MFCD04967153 CAS Number: 1002032-57-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...