Ethyl-3-nitro-1-propyl-1H-pyrazole-5-carboxylate

Supplier: Matrix Scientific

080350-500MG 080350-1G
102669-230EA 516.92 USD
102669-230 102669-232
Ethyl-3-nitro-1-propyl-1H-pyrazole-5-carboxylate
Ethyl-3-nitro-1-propyl-1H-pyrazole-5-carboxylate

Hazard: Irritant

Formula: C₉H₁₃N₃O₄
MDL Number: MFCD10001567
CAS Number: 1170291-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR