Methyl (4-chloro-5-methyl-3-nitro-1H-pyrazol-1-yl)acetate
Supplier: Matrix Scientific
080349-500MG
080349-1G
102669-226EA
516.92
USD
102669-226
102669-228
Methyl (4-chloro-5-methyl-3-nitro-1H-pyrazol-1-yl)acetate
Methyl (4-chloro-5-methyl-3-nitro-1H-pyrazol-1-yl)acetate
Hazard: Irritant
Formula:
C₇H₈ClN₃O₄ |
MDL Number:
MFCD09475384 CAS Number: 1171645-95-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...