Methyl-1-ethyl-4-nitro-1H-pyrazole-3-carboxylate

Supplier: Matrix Scientific

Synonyms: Methyl 1-ethyl-4-nitro-1H-pyrazole-3-carboxylate

080348-500MG 080348-1G
102669-222EA 516.92 USD
102669-222 102669-224
Methyl-1-ethyl-4-nitro-1H-pyrazole-3-carboxylate
Methyl-1-ethyl-4-nitro-1H-pyrazole-3-carboxylate

Hazard: Irritant

Formula: C₇H₉N₃O₄
MW: 199.17 g/mol
MDL Number: MFCD09475383
CAS Number: 923283-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR