Ethyl-3-(3-methoxy-4-nitro-1H-pyrazol-1-yl)propanoate

Supplier: Matrix Scientific

080347-500MG 080347-1G
102669-218EA 516.92 USD
102669-218 102669-220
Ethyl-3-(3-methoxy-4-nitro-1H-pyrazol-1-yl)propanoate
Ethyl-3-(3-methoxy-4-nitro-1H-pyrazol-1-yl)propanoate

Hazard: Irritant

Formula: C₉H₁₃N₃O₅
MDL Number: MFCD04969821
CAS Number: 1001500-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR