Ethyl-2-(4-nitro-1H-pyrazol-1-yl)butanoate

Supplier: Matrix Scientific

080346-500MG 080346-1G
102669-214EA 516.92 USD
102669-214 102669-216
Ethyl-2-(4-nitro-1H-pyrazol-1-yl)butanoate
Ethyl-2-(4-nitro-1H-pyrazol-1-yl)butanoate

Hazard: Irritant

Formula: C₉H₁₃N₃O₄
MDL Number: MFCD04969812
CAS Number: 1005668-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR