Ethyl-2-(5-methyl-3-nitro-1H-pyrazol-1-yl)butanoate

Supplier: Matrix Scientific

080345-500MG 080345-1G
102669-210EA 516.92 USD
102669-210 102669-212
Ethyl-2-(5-methyl-3-nitro-1H-pyrazol-1-yl)butanoate
Ethyl-2-(5-methyl-3-nitro-1H-pyrazol-1-yl)butanoate

Hazard: Irritant

Formula: C₁₀H₁₅N₃O₄
MDL Number: MFCD04969788
CAS Number: 1005576-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR