Ethyl (5-methyl-3-nitro-1H-pyrazol-1-yl)acetate

Supplier: Matrix Scientific

080340-500MG 080340-1G
102669-188EA 516.92 USD
102669-188 102669-190
Ethyl (5-methyl-3-nitro-1H-pyrazol-1-yl)acetate
Ethyl (5-methyl-3-nitro-1H-pyrazol-1-yl)acetate

Hazard: Irritant

Formula: C₈H₁₁N₃O₄
MDL Number: MFCD00682562
CAS Number: 321165-21-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR