Ethyl-4-(5-methyl-3-nitro-1H-pyrazol-1-yl)butanoate

Supplier: Matrix Scientific

080337-500MG 080337-1G
102669-176EA 387.69 USD
102669-176 102669-178
Ethyl-4-(5-methyl-3-nitro-1H-pyrazol-1-yl)butanoate
Ethyl-4-(5-methyl-3-nitro-1H-pyrazol-1-yl)butanoate

Hazard: Irritant

Formula: C₁₀H₁₅N₃O₄
MDL Number: MFCD06167036
CAS Number: 898053-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR