[1-(1,3-Dimethyl-1H-pyrazol-4-yl)propyl]amine

Supplier: Matrix Scientific

080335-500MG 080335-1G 080335-5G
102669-166EA 646.15 USD
102669-166 102669-168 102669-170
[1-(1,3-Dimethyl-1H-pyrazol-4-yl)propyl]amine
[1-(1,3-Dimethyl-1H-pyrazol-4-yl)propyl]amine

Hazard: Irritant

Formula: C₈H₁₅N₃
MDL Number: MFCD08700793
CAS Number: 1007501-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR