[2-(3-Methyl-1H-pyrazol-1-yl)propyl]amine
Supplier: Matrix Scientific
080333-500MG
080333-1G
080333-5G
102669-154EA
646.15
USD
102669-154
102669-156
102669-158
[2-(3-Methyl-1H-pyrazol-1-yl)propyl]amine
[2-(3-Methyl-1H-pyrazol-1-yl)propyl]amine
Hazard: Irritant
Formula:
C₇H₁₃N₃ |
MDL Number:
MFCD11215315 CAS Number: 1172803-90-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...