{1-Methyl-2-[5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]ethyl}amine
Supplier: Matrix Scientific
080331-500MG
080331-1G
080331-5G
102669-142EA
516.92
USD
102669-142
102669-144
102669-146
{1-Methyl-2-[5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]ethyl}amine
{1-Methyl-2-[5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]ethyl}amine
Hazard: Irritant
Formula:
C₈H₁₂F₃N₃ |
MDL Number:
MFCD04970169 CAS Number: 1006483-37-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...