[2-(4-Chloro-3,5-dimethyl-1H-pyrazol-1-yl)-3-methylbutyl]amine

Supplier: Matrix Scientific

080330-500MG 080330-1G 080330-5G
102669-136EA 516.92 USD
102669-136 102669-138 102669-140
[2-(4-Chloro-3,5-dimethyl-1H-pyrazol-1-yl)-3-methylbutyl]amine
[2-(4-Chloro-3,5-dimethyl-1H-pyrazol-1-yl)-3-methylbutyl]amine

Hazard: Irritant

Formula: C₁₀H₁₈ClN₃
MDL Number: MFCD10001527
CAS Number: 1171866-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR