1-(2,2-Difluoroethyl)-3-methyl-1H-pyrazol-5-amine
Supplier: Matrix Scientific
080322-500MG
080322-1G
080322-5G
102669-088EA
646.15
USD
102669-088
102669-090
102669-092
1-(2,2-Difluoroethyl)-3-methyl-1H-pyrazol-5-amine
1-(2,2-Difluoroethyl)-3-methyl-1H-pyrazol-5-amine
Hazard: Irritant
Formula:
C₆H₉F₂N₃ |
MDL Number:
MFCD09473470 CAS Number: 1172884-50-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...