{3-[1',5'-Dimethyl-5-(trifluoromethyl)-1'H,2H-3,4'-bipyrazol-2-yl]propyl}amine

Supplier: Matrix Scientific

080320-500MG 080320-1G 080320-5G
102669-076EA 387.69 USD
102669-076 102669-078 102669-080
{3-[1',5'-Dimethyl-5-(trifluoromethyl)-1'H,2H-3,4'-bipyrazol-2-yl]propyl}amine
{3-[1',5'-Dimethyl-5-(trifluoromethyl)-1'H,2H-3,4'-bipyrazol-2-yl]propyl}amine

Hazard: Irritant

Formula: C₁₂H₁₆F₃N₅
MDL Number: MFCD09473382
CAS Number: 1171853-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR