{3-[1',5'-Dimethyl-5-(trifluoromethyl)-1H,1'H-3,4'-bipyrazol-1-yl]propyl}amine

Supplier: Matrix Scientific

080319-500MG 080319-1G 080319-5G
102669-070EA 387.69 USD
102669-070 102669-072 102669-074
{3-[1',5'-Dimethyl-5-(trifluoromethyl)-1H,1'H-3,4'-bipyrazol-1-yl]propyl}amine
{3-[1',5'-Dimethyl-5-(trifluoromethyl)-1H,1'H-3,4'-bipyrazol-1-yl]propyl}amine

Hazard: Irritant

Formula: C₁₂H₁₆F₃N₅
MDL Number: MFCD09473359
CAS Number: 1170015-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR