3-(4-Amino-5-mercapto-4H-1,2,4-triazol-3-yl)propan-1-ol
Supplier: Matrix Scientific
080318-500MG
080318-1G
080318-5G
102669-064EA
516.92
USD
102669-064
102669-066
102669-068
3-(4-Amino-5-mercapto-4H-1,2,4-triazol-3-yl)propan-1-ol
3-(4-Amino-5-mercapto-4H-1,2,4-triazol-3-yl)propan-1-ol
Hazard: Irritant
Formula:
C₅H₁₀N₄OS |
MDL Number:
MFCD08559006 CAS Number: 828295-64-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...