1-(2,2-Difluoroethyl)-1H-pyrazol-4-amine hydrochloride
Supplier: Matrix Scientific
080317-500MG
080317-1G
080317-5G
102669-058EA
553.85
USD
102669-058
102669-060
102669-062
1-(2,2-Difluoroethyl)-1H-pyrazol-4-amine hydrochloride
1-(2,2-Difluoroethyl)-1H-pyrazol-4-amine hydrochloride
Hazard: Irritant
Formula:
C₅H₈ClF₂N₃ |
MDL Number:
MFCD15142911 CAS Number: 1197231-23-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...