1-Ethyl-1,6-dihydro-7H-pyrazolo[4,3-d]pyrimidin-7-one

Supplier: Matrix Scientific

080314-500MG 080314-1G 080314-5G
102669-040EA 516.92 USD
102669-040 102669-042 102669-044
1-Ethyl-1,6-dihydro-7H-pyrazolo[4,3-d]pyrimidin-7-one
1-Ethyl-1,6-dihydro-7H-pyrazolo[4,3-d]pyrimidin-7-one

Hazard: Irritant

Formula: C₇H₈N₄O
MDL Number: MFCD04050119
CAS Number: 674788-53-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR