[3-(4-Chloro-3,5-dimethyl-1H-pyrazol-1-yl)butyl]amine

Supplier: Matrix Scientific

080312-500MG 080312-1G 080312-5G
102669-028EA 516.92 USD
102669-028 102669-030 102669-032
[3-(4-Chloro-3,5-dimethyl-1H-pyrazol-1-yl)butyl]amine
[3-(4-Chloro-3,5-dimethyl-1H-pyrazol-1-yl)butyl]amine

Hazard: Irritant

Formula: C₉H₁₆ClN₃
MDL Number: MFCD06805044
CAS Number: 1006480-96-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR