[3-(4-Chloro-1H-pyrazol-1-yl)pentyl]amine
Supplier: Matrix Scientific
080311-500MG
080311-1G
080311-5G
102669-022EA
516.92
USD
102669-022
102669-024
102669-026
[3-(4-Chloro-1H-pyrazol-1-yl)pentyl]amine
[3-(4-Chloro-1H-pyrazol-1-yl)pentyl]amine
Hazard: Irritant
Formula:
C₈H₁₄ClN₃ |
MDL Number:
MFCD06805038 CAS Number: 1006480-90-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...