[3-(1H-Pyrazol-1-yl)pentyl]amine

Supplier: Matrix Scientific

080310-500MG 080310-1G 080310-5G
102669-016EA 516.92 USD
102669-016 102669-018 102669-020
[3-(1H-Pyrazol-1-yl)pentyl]amine
[3-(1H-Pyrazol-1-yl)pentyl]amine

Hazard: Irritant

Formula: C₈H₁₅N₃
MDL Number: MFCD06805022
CAS Number: 1006334-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR