[2-(3-Ethoxy-4-nitro-1H-pyrazol-1-yl)ethyl]amine hydrochloride

Supplier: Matrix Scientific

080309-500MG 080309-1G 080309-5G
102669-010EA 516.92 USD
102669-010 102669-012 102669-014
[2-(3-Ethoxy-4-nitro-1H-pyrazol-1-yl)ethyl]amine hydrochloride
[2-(3-Ethoxy-4-nitro-1H-pyrazol-1-yl)ethyl]amine hydrochloride

Hazard: Irritant

Formula: C₇H₁₃ClN₄O₃
MDL Number: MFCD22421974
CAS Number: 1417570-01-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR