Ethyl-2-(4-amino-1H-pyrazol-1-yl)propanoate hydrochloride
Supplier: Matrix Scientific
080307-500MG
080307-1G
080307-5G
102668-998EA
646.15
USD
102668-998
102669-000
102669-002
Ethyl-2-(4-amino-1H-pyrazol-1-yl)propanoate hydrochloride
Ethyl-2-(4-amino-1H-pyrazol-1-yl)propanoate hydrochloride
Hazard: Irritant
Formula:
C₈H₁₄ClN₃O₂ |
MDL Number:
MFCD22421973 CAS Number: 1417567-78-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...