[3-(3-Methoxy-4-nitro-1H-pyrazol-1-yl)propyl]amine hydrochloride
Supplier: Matrix Scientific
080304-500MG
080304-1G
080304-5G
102668-980EA
516.92
USD
102668-980
102668-982
102668-984
[3-(3-Methoxy-4-nitro-1H-pyrazol-1-yl)propyl]amine hydrochloride
[3-(3-Methoxy-4-nitro-1H-pyrazol-1-yl)propyl]amine hydrochloride
Hazard: Irritant
Formula:
C₇H₁₃ClN₄O₃ |
MDL Number:
MFCD22421970 CAS Number: 1417567-75-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...