[2-(3-Methoxy-4-nitro-1H-pyrazol-1-yl)ethyl]amine hydrochloride

Supplier: Matrix Scientific

080303-500MG 080303-1G 080303-5G
102668-974EA 516.92 USD
102668-974 102668-976 102668-978
[2-(3-Methoxy-4-nitro-1H-pyrazol-1-yl)ethyl]amine hydrochloride
[2-(3-Methoxy-4-nitro-1H-pyrazol-1-yl)ethyl]amine hydrochloride

Hazard: Irritant

Formula: C₆H₁₁ClN₄O₃
MDL Number: MFCD22421969
CAS Number: 1417569-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR