[3-(4-Chloro-1H-pyrazol-1-yl)-2-methylpropyl]amine

Supplier: Matrix Scientific

080302-500MG 080302-1G 080302-5G
102668-968EA 516.92 USD
102668-968 102668-970 102668-972
[3-(4-Chloro-1H-pyrazol-1-yl)-2-methylpropyl]amine
[3-(4-Chloro-1H-pyrazol-1-yl)-2-methylpropyl]amine

Hazard: Irritant

Formula: C₇H₁₂ClN₃
MDL Number: MFCD06805027
CAS Number: 1006319-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR