[2-Methyl-3-(3-methyl-1H-pyrazol-1-yl)propyl]amine

Supplier: Matrix Scientific

080299-500MG 080299-1G 080299-5G
102668-950EA 646.15 USD
102668-950 102668-952 102668-954
[2-Methyl-3-(3-methyl-1H-pyrazol-1-yl)propyl]amine
[2-Methyl-3-(3-methyl-1H-pyrazol-1-yl)propyl]amine

Hazard: Irritant

Formula: C₈H₁₅N₃
MDL Number: MFCD06805016
CAS Number: 1006319-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR