3-Ethyl-5-(ethylthio)-4H-1,2,4-triazol-4-amine

Supplier: Matrix Scientific

080294-500MG 080294-5G
102668-920EA 516.92 USD
102668-920 102668-924
3-Ethyl-5-(ethylthio)-4H-1,2,4-triazol-4-amine
3-Ethyl-5-(ethylthio)-4H-1,2,4-triazol-4-amine

Hazard: Irritant

Formula: C₆H₁₂N₄S
MDL Number: MFCD03536363
CAS Number: 449745-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR