3-(Ethylthio)-5-(2-furyl)-4H-1,2,4-triazol-4-amine
Supplier: Matrix Scientific
080288-500MG
080288-5G
102668-884EA
387.69
USD
102668-884
102668-888
3-(Ethylthio)-5-(2-furyl)-4H-1,2,4-triazol-4-amine
3-(Ethylthio)-5-(2-furyl)-4H-1,2,4-triazol-4-amine
Hazard: Irritant
Formula:
C₈H₁₀N₄OS |
MDL Number:
MFCD03532486 CAS Number: 865593-32-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...