3-(Difluoromethyl)-5-(ethylthio)-4H-1,2,4-triazol-4-amine
Supplier: Matrix Scientific
080286-500MG
080286-5G
102668-872EA
516.92
USD
102668-872
102668-876
3-(Difluoromethyl)-5-(ethylthio)-4H-1,2,4-triazol-4-amine
3-(Difluoromethyl)-5-(ethylthio)-4H-1,2,4-triazol-4-amine
Hazard: Irritant
Formula:
C₅H₈F₂N₄S |
MDL Number:
MFCD04969120 CAS Number: 832737-84-5 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...