[(1-Ethyl-1H-pyrazol-5-yl)methyl]methylamine
Supplier: Matrix Scientific
080284-500MG
080284-5G
102668-860EA
646.15
USD
102668-860
102668-864
[(1-Ethyl-1H-pyrazol-5-yl)methyl]methylamine
[(1-Ethyl-1H-pyrazol-5-yl)methyl]methylamine
Hazard: Irritant
Formula:
C₇H₁₃N₃ |
MDL Number:
MFCD03419814 CAS Number: 1002651-67-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...