4-Amino-5-(difluoromethyl)-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

080280-500MG 080280-5G
102668-836EA 646.15 USD
102668-836 102668-840
4-Amino-5-(difluoromethyl)-4H-1,2,4-triazole-3-thiol
4-Amino-5-(difluoromethyl)-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₃H₄F₂N₄S
MDL Number: MFCD03423399
CAS Number: 832740-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR