5-(Chloromethyl)-1-ethylpyrazolo[1,5-a]pyrimidin-7(1H)-one

Supplier: Matrix Scientific

080274-500MG 080274-5G
102668-800EA 646.15 USD
102668-800 102668-804
5-(Chloromethyl)-1-ethylpyrazolo[1,5-a]pyrimidin-7(1H)-one
5-(Chloromethyl)-1-ethylpyrazolo[1,5-a]pyrimidin-7(1H)-one

Hazard: Irritant

Formula: C₉H₁₀ClN₃O
MW: 211.65 g/mol
MDL Number: MFCD09859224
CAS Number: 1018165-90-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR