1-[4-Methyl-6-(trifluoromethyl)pyrimidin-2-yl]piperidine-3-carboxylic acid

Supplier: Matrix Scientific

080272-500MG 080272-5G
102668-788EA 387.69 USD
102668-788 102668-792
1-[4-Methyl-6-(trifluoromethyl)pyrimidin-2-yl]piperidine-3-carboxylic acid
1-[4-Methyl-6-(trifluoromethyl)pyrimidin-2-yl]piperidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₄F₃N₃O₂
MDL Number: MFCD05861935
CAS Number: 861451-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR