7-(1,3,5-Trimethyl-1H-pyrazol-4-yl)pyrazolo[1,5-a]pyrimidine-2-carboxylic acid
Supplier: Matrix Scientific
080262-500MG
080262-5G
102668-728EA
516.92
USD
102668-728
102668-732
7-(1,3,5-Trimethyl-1H-pyrazol-4-yl)pyrazolo[1,5-a]pyrimidine-2-carboxylic acid
7-(1,3,5-Trimethyl-1H-pyrazol-4-yl)pyrazolo[1,5-a]pyrimidine-2-carboxylic acid
Hazard: Irritant
Formula:
C₁₃H₁₃N₅O₂ MW: 271.28 g/mol |
MDL Number:
MFCD06739335 CAS Number: 1004727-47-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...