3-(3,5-Dimethyl-4-nitro-1H-pyrazol-1-yl)-2-methylpropanoic acid
Supplier: Matrix Scientific
080256-500MG
080256-5G
102668-692EA
516.92
USD
102668-692
102668-696
3-(3,5-Dimethyl-4-nitro-1H-pyrazol-1-yl)-2-methylpropanoic acid
3-(3,5-Dimethyl-4-nitro-1H-pyrazol-1-yl)-2-methylpropanoic acid
Hazard: Irritant
Formula:
C₉H₁₃N₃O₄ |
MDL Number:
MFCD04967183 CAS Number: 1005640-74-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...