1-(Carboxymethyl)piperidine-4-carboxylic acid

Supplier: Matrix Scientific

080252-500MG 080252-5G
102668-668EA 516.92 USD
102668-668 102668-672
1-(Carboxymethyl)piperidine-4-carboxylic acid
1-(Carboxymethyl)piperidine-4-carboxylic acid

Hazard: Irritant

Formula: C₈H₁₃NO₄
MDL Number: MFCD00474246
CAS Number: 53919-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR