2-[4-Chloro-3-(difluoromethyl)-5-methyl-1H-pyrazol-1-yl]propanoic acid

Supplier: Matrix Scientific

080246-500MG 080246-5G
102668-634EA 516.92 USD
102668-634 102668-638
2-[4-Chloro-3-(difluoromethyl)-5-methyl-1H-pyrazol-1-yl]propanoic acid
2-[4-Chloro-3-(difluoromethyl)-5-methyl-1H-pyrazol-1-yl]propanoic acid

Hazard: Irritant

Formula: C₈H₉ClF₂N₂O₂
MDL Number: MFCD03419700
CAS Number: 1005584-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR