3-(4,5-Dichloro-3-methyl-1H-pyrazol-1-yl)propanenitrile

Supplier: Matrix Scientific

080243-500MG 080243-5G
102668-618EA 516.92 USD
102668-618 102668-622
3-(4,5-Dichloro-3-methyl-1H-pyrazol-1-yl)propanenitrile
3-(4,5-Dichloro-3-methyl-1H-pyrazol-1-yl)propanenitrile

Hazard: Irritant

Formula: C₇H₇Cl₂N₃
MDL Number: MFCD04969828
CAS Number: 1001500-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR