Oxo[(5-propyl-1,3,4-thiadiazol-2-yl)amino]acetic acid

Supplier: Matrix Scientific

080226-500MG 080226-5G
102668-524EA 387.69 USD
102668-524 102668-528
Oxo[(5-propyl-1,3,4-thiadiazol-2-yl)amino]acetic acid
Oxo[(5-propyl-1,3,4-thiadiazol-2-yl)amino]acetic acid

Hazard: Irritant

Formula: C₇H₉N₃O₃S
MDL Number: MFCD15181831
CAS Number: 83244-83-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR